missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetone, 95%, stab. with 0.1% epoxidized soybean oil

Product Code. 11471890
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471890 250 g 250g
11490980 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471890 Supplier Thermo Scientific Alfa Aesar Supplier No. A11922.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol

Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetone was used in the synthesis of double-chain nonionic surfactants with an acid decomposition function via acid-catalyzed condensation with fatty alcohols (octyl, decyl and dodecyl). It was also used in the synthesis of meso-tetramethyl tetrakis-(4-phenoxy acetone)calix[4]pyrrole. It can also be used to make dye couplers for color photography and used in the Feist-Benary synthesis of furans.

Solubility
Soluble in water (124g/L), ether and chloroform.

Notes
Refrigerate. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Do not store together with strong basic or oxidizing materials.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
SMILES CC(=O)CCl

Specifications

Melting Point °C to 45°C
Density 1.161
Boiling Point 118°C to 120°C
Flash Point 40°C (104°F)
Assay Percent Range 95%, Stabilized with 0.1% Epoxidized Soybean Oil
Linear Formula CH2COCH3
Refractive Index 1.434
Quantity 250 g
UN Number UN1695
Beilstein 605369
Merck Index 14,2114
Synonym 1-propanol, 3-chloro-, acetate, 3-chloropropanol acetate, 3-chlorprop-1-ylacetat, gamma-chloropropyl acetate, 3-chlorprop-1-ylacetat german, acetic acid 3-chloro-propyl ester, 1-acetoxy-3-chloropropane, .gamma.-chloropropyl acetate, 3-chloro-propyl acetate, acetic acid 3-chloropropyl ester
Solubility Information Soluble in water (124g/L),ether and chloroform.
IUPAC Name 3-chloropropyl acetate
PubChem CID 12334
Formula Weight 92.53
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Chloroacetone, Stabilized with 0.1% epoxidized soybean oil
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.