missing translation for 'onlineSavingsMsg'
Learn More

Chloroacetyl Anhydride, TRC

Product Code. 30353662
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353662 10 g -
30366460 1 g 1g
30350733 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353662 Supplier Toronto Research Chemicals Supplier No. TRCC32720510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-88-8
Molecular Formula C4H4Cl2O3
Molecular Weight (g/mol) 170.98
Synonym Bischloroacetic Acid Anhydride, Chloroacetic Acid Anhydride, Chloroacetic Anhydride, alpha,alpha'-Dichloroacetic Anhydride, NSC 71207
IUPAC Name (2-chloroacetyl) 2-chloroacetate
SMILES O=C(OC(=O)CCl)CCl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H4Cl2O3/c5-1-3(7)9-4(8)2-6/h1-2H2
Formula Weight 169.9537
Chemical Name or Material Chloroacetyl Anhydride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.