missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ≥99.5%, ACS reagent

Product Code. 10461011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
Unit Size:
1kg
2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10461011 1 kg EcoSafPak™ Glass Bottle 1kg
10484115 2.5 kg Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10461011 Supplier Thermo Scientific Acros Supplier No. 404490010
  • ONLINE EXCLUSIVE PRICE
    83.45 €
    81.00 € / 1kg
    Save 2.45 €
    2% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Density 1.1075g/mL
Quantity 1 kg
Infrared Spectrum Authentic
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.5230 to 1.5250 (20°C, 589nm)
Residue after Evaporation 0.02% max.
Assay Percent Range 99.6%
Titratable Acid 0.004 meq/g max.
Linear Formula C6H5Cl
Vapor Pressure Vapor Pressure: 12mbar at 20°C
UN Number 1134
Beilstein 05, 199
Merck Index 15, 2122
Formula Weight 112.56
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Chlorobenzene, For analysis ACS
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.