missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, 99%

Product Code. 11470147
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470147 2500 mL 2500mL
11460147 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470147 Supplier Thermo Scientific Alfa Aesar Supplier No. B21052.0F
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

Solubility
Miscible with alcohol, chloroform, diethyl ether and benzene. Slightly immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Density 1.106
Quantity 2500 mL
Odor Aromatic
Refractive Index 1.524
UN Number UN1134
Beilstein 605632
Merck Index 14,2121
Boiling Point 132°C
Solubility Information Miscible with alcohol,chloroform,diethyl ether and benzene. Slightly immiscible with water.
Formula Weight 112.56
Percent Purity 99%
Chemical Name or Material Chlorobenzene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Chlorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.