missing translation for 'onlineSavingsMsg'
Learn More

Chlorobenzene, ACS, 99.5%

Product Code. 11327868
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327868 4 x 1 L 4 L
11455170 500 mL 500mL
11337868 1 L 1 L
11347868 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11327868 Supplier Thermo Scientific Alfa Aesar Supplier No. 036401.M1
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-90-7 | C6H5Cl | 112.556 g/mol

Chlorobenzene is used in organic synthesis and dyestuffs. It is also used as a solvent for adhesives, drugs, rubber, paints and dry cleaning, and as a fiber-swelling agent in textile processing. It is utilized in chemoenzymatic synthesis of (+)-aspicilin and cyclohexane by hydrogenation over colloidal Pt nanocatalysts under ambient conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Color Colorless
Density 1.106 g/mL
Boiling Point 132°C
Quantity 4 x 1 L
Flash Point 28°C (82°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.524
UN Number UN1134
Beilstein 605632
Merck Index 14,2121
Solubility Information Miscible with methanol and diethyl ether. Immiscible with water.
Formula Weight 112.56
Percent Purity 99.5%
Grade ACS
Chemical Name or Material Chlorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.