missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(4-chlorophenyl)phosphine, 97%

Product Code. 11455630
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455630 1 g 1g
11465630 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455630 Supplier Thermo Scientific Alfa Aesar Supplier No. H25886.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13685-26-2 | C12H8Cl3P | 289.52 g/mol

Chlorobis(4-chlorophenyl)phosphine are now employed in tuning the properties of catalyst. Also used as primary and secondary intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(4-chlorophenyl)phosphine are now employed in tuning the properties of catalyst. Also used as primary and secondary intermediates.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13685-26-2
Molecular Formula C12H8Cl3P
Molecular Weight (g/mol) 289.52
MDL Number MFCD09909564
InChI Key UISLTICQIVPVGS-UHFFFAOYSA-N
Synonym chlorobis 4-chlorophenyl phosphine, chlorobis 4-chlorophenyl phosphane, bis 4-chlorophenyl chlorophosphine, chloro-bis 4-chlorophenyl phosphane, bis 4-chlorophenyl phosphinous chloride, chlorobis 4-chlorophenyl phosphine,98+%
PubChem CID 18705966
IUPAC Name chloro-bis(4-chlorophenyl)phosphane
SMILES C1=CC(=CC=C1P(C2=CC=C(C=C2)Cl)Cl)Cl

Specifications

Melting Point 52°C to 53°C
Boiling Point 184°C to 186°C (2.5 mmHg)
Quantity 1 g
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 289.53
Percent Purity 97%
Chemical Name or Material Chlorobis(4-chlorophenyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.