missing translation for 'onlineSavingsMsg'
Learn More

Chlorobis(ethylene)rhodium(I) Dimer, TRC

Product Code. 30179798
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179798 500 mg 500mg
30180431 100 mg 100mg
30180298 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179798 Supplier Toronto Research Chemicals Supplier No. TRCC984708500MG
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12081-16-2
Molecular Formula C8H16Cl2Rh2
Molecular Weight (g/mol) 388.93
Synonym Di-μ-chlorotetrakis(η2-ethene)dirhodium, Bis((μ-chloro)bis(ethylene)rhodium);
IUPAC Name Rhodium, di-μ-chlorotetrakis(η2-ethene)di- (9CI, ACI)
SMILES C=C.C=C.[][Rh](Cl1)([])Cl[Rh]1([])[].C=C.C=C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/4C2H4.2ClH.2Rh/c4*1-2;;;;/h4*1-2H2;2*1H;;/q;;;;;;2*+1/p-2
Formula Weight 387.87
Chemical Name or Material Chlorobis(ethylene)rhodium(I) Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.