missing translation for 'onlineSavingsMsg'
Learn More

Chlorocyclohexane, TRC

Product Code. 30358602
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358602 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358602 Supplier Toronto Research Chemicals Supplier No. TRCC32722550G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorocyclohexane
CAS 542-18-7
Molecular Formula C6H11Cl
Quantity 50 g
Synonym Monochlorocyclohexane, Cyclohexyl Chloride, NSC 8434
InChI Formula InChI=1S/C6H11Cl/c7-6-4-2-1-3-5-6/h6H,1-5H2
SMILES ClC1CCCCC1
IUPAC Name chlorocyclohexane
Molecular Weight (g/mol) 118.6
Formula Weight 118.0549
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.