missing translation for 'onlineSavingsMsg'
Learn More

Chlorodicyclohexylphosphine, 97%

Product Code. 10324402 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10324402 1 g Glass bottle 1g
10008851 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10324402 Supplier Thermo Scientific Acros Supplier No. 353290010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 16523-54-9 | C12H22ClP | 232.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16523-54-9
Molecular Formula C12H22ClP
Molecular Weight (g/mol) 232.73
MDL Number MFCD00014294
InChI Key AKJFBIZAEPTXIL-UHFFFAOYSA-N
Synonym chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
PubChem CID 2734135
IUPAC Name chloro(dicyclohexyl)phosphane
SMILES ClP(C1CCCCC1)C1CCCCC1

Specifications

Color Colorless
Density 1.0540g/mL
Boiling Point 165.0°C (12.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula (C6H11)2PCl
Refractive Index 1.5310 to 1.5340
Quantity 1 g
Specific Gravity 1.054
Solubility Information Solubility in water: reacts violently with water
Formula Weight 232.74
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chlorodicyclohexylphosphine
Show More Show Less
compliance-icons
Product Identifier
  • Chlorodicyclohexylphosphine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.