missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiethylisopropylsilane, 95+%, Thermo Scientific™

Product Code. 11975961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11975961 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11975961 Supplier Thermo Scientific Acros Supplier No. 435650250
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 107149-56-4
Color Colorless
Density 0.9000g/mL
Boiling Point 75.0°C
Flash Point 38°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C7H17ClSi
Linear Formula (CH3)2CHSi(C2H5)2Cl
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
MDL Number MFCD00191631
Quantity 25 mL
Specific Gravity 0.9
Synonym chlorodiethylisopropylsilane, diethylisopropylsilyl chloride, acmc-1bql9, isopropyldiethylchlorosilane, silane,chlorodiethyl 1-methylethyl, silane, chlorodiethyl 1-methylethyl
Solubility Information Solubility in water: reacts
InChI Key OSBPGFIPLLCQMO-UHFFFAOYSA-N
SMILES CC[Si](Cl)(CC)C(C)C
IUPAC Name chloro-diethyl-propan-2-ylsilane
Molecular Weight (g/mol) 164.75
PubChem CID 4486980
Formula Weight 164.75
Percent Purity 95+%
Physical Form Liquid
Chemical Name or Material chlorodiethylisopropylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorodiethylisopropylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
  • H226-Flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.