missing translation for 'onlineSavingsMsg'
Learn More

Chlorodifluoro-acetic Acid Sodium Salt, TRC

Product Code. 30360121
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30360121 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360121 Supplier Toronto Research Chemicals Supplier No. TRCC36546050G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorodifluoro-acetic Acid Sodium Salt
CAS 1895-39-2
Molecular Formula C2ClF2NaO2
Quantity 50 g
Synonym 2-Chloro-2,2-difluoro-acetic Acid Sodium Salt, Chlorodifluoroacetic Acid Sodium Salt, Sodium Chlorodifluoroacetate, Sodium Difluorochloroacetate
InChI Formula InChI=1S/C2HClF2O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1
SMILES [Na+].[O-]C(=O)C(F)(F)Cl
IUPAC Name sodium;2-chloro-2,2-difluoroacetate
Molecular Weight (g/mol) 152.46
Formula Weight 151.9453
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.