missing translation for 'onlineSavingsMsg'
Learn More

chlorodifluoroacetic acid sodium salt, 96%

Product Code. 10711252 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711252 25 g Glass bottle 25g
10494115 5 g Glass bottle 5g
10390464 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10711252 Supplier Thermo Scientific Acros Supplier No. 404590250
  • ONLINE EXCLUSIVE PRICE
    143.00 €
    138.00 € / 25g
    Save 5.00 €
    3% Off
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1895-39-2 | C2ClF2NaO2 | 152.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1895-39-2
Molecular Formula C2ClF2NaO2
Molecular Weight (g/mol) 152.46
MDL Number MFCD00064771
InChI Key MRTAVLDNYYEJHK-UHFFFAOYSA-M
Synonym sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
PubChem CID 2734985
IUPAC Name sodium;2-chloro-2,2-difluoroacetate
SMILES C(=O)(C(F)(F)Cl)[O-].[Na+]

Specifications

Melting Point 195.0°C to 201.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula ClF2CCO2Na
Quantity 25 g
Fieser 01,1227; 02,379; 03,265; 05,603
Formula Weight 152.46
Percent Purity 96%
Physical Form Powder
Chemical Name or Material chlorodifluoroacetic acid sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Chlorodifluoroacetic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.