missing translation for 'onlineSavingsMsg'
Learn More

chlorodiisopropylsilane, 95%

Product Code. 11925841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11925841 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11925841 Supplier Thermo Scientific Acros Supplier No. 313440050
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2227-29-4 | C6H14ClSi | 149.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2227-29-4
Color Colorless
Density 0.8720g/mL
Boiling Point 137.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14ClSi
Refractive Index 1.4280 to 1.4300 (20°C, 589nm)
MDL Number MFCD00054896
Quantity 5 g
Merck Index 15,2136
Specific Gravity 0.872
Synonym chlorodiisopropylsilane, diisopropylchlorosilane, unii-dvg177547j, silane, chloro diisopropyl, zlchem 966, chlorodiisopropylsilyl, chlorobis propan-2-yl silane, chloro-di propan-2-yl silicon
Solubility Information Solubility in water: reacts
InChI Key IGSUJBNDAWQLST-UHFFFAOYSA-N
SMILES CC(C)[Si](Cl)C(C)C
IUPAC Name chloro-di(propan-2-yl)silicon
Molecular Weight (g/mol) 149.71
PubChem CID 6365034
Formula Weight 150.73
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material chlorodiisopropylsilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diisopropylchlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diisopropylchlorosilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.