missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethyl-3,3,3-trifluoropropylsilane, 96%, Thermo Scientific™

Product Code. 11389305
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389305 100 g 100g
11369305 5 g 5g
11379305 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389305 Supplier Thermo Scientific Alfa Aesar Supplier No. L16666.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1481-41-0
Molecular Formula C5H10ClF3Si
Molecular Weight (g/mol) 190.665
MDL Number MFCD00053211
InChI Key KBAZUXSLKGQRJF-UHFFFAOYSA-N
Synonym trifluoropropyldimethylchlorosilane, chlorodimethyl-3,3,3-trifluoropropylsilane, 3,3,3-trifluoropropyl dimethylchlorosilane, chlorodimethyl 3,3,3-trifluoropropyl silane, silane, chlorodimethyl 3,3,3-trifluoropropyl, dimethyl 3,3,3-trifluoropropyl chlorosilane, chloro-dimethyl-3,3,3-trifluoropropyl silane, acmc-1c1aj, chloro dimethyl 3,3,3-trifluoropropyl silane, 3,3,3-trifluoropropylchlorodimethylsilane
PubChem CID 73866
IUPAC Name chloro-dimethyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](C)(CCC(F)(F)F)Cl

Specifications

Boiling Point 118°C to 120°C
Quantity 100 g
UN Number UN2985
Beilstein 1741726
Flash Point 20°C (68°F)
Formula Weight 190.67
Percent Purity 96%
Refractive Index 1.375
Sensitivity Moisture sensitive
Density 1.105
Chemical Name or Material Chlorodimethyl-3,3,3-trifluoropropylsilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.