missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylsilane, 97%

Product Code. 10387290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387290 10 g Glass bottle 10g
10541611 50 g Glass bottle 50g
10711935 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10387290 Supplier Thermo Scientific Acros Supplier No. 162840100
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1066-35-9 | C2H6ClSi | 93.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-35-9
Molecular Formula C2H6ClSi
Molecular Weight (g/mol) 93.61
MDL Number MFCD00000495
InChI Key QABCGOSYZHCPGN-UHFFFAOYSA-N
Synonym chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical
PubChem CID 6327132
IUPAC Name chloro(dimethyl)silicon
SMILES C[Si](C)Cl

Specifications

Melting Point -111°C
Color Colorless
Density 0.8500g/mL
Boiling Point 34.6°C
Flash Point -28°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula (CH3)2SiHCl
Refractive Index 1.3820 to 1.384
Quantity 10 g
Fieser 13,73
Specific Gravity 0.85
Solubility Information Solubility in water: reacts
Formula Weight 94.62
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material chlorodimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.