missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylsilane, 97%

Product Code. 15433067
Change view
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Unit Size:
10mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15433067 10 mL 10mL
15443067 50 mL 50mL
15453067 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15433067 Supplier Thermo Scientific Alfa Aesar Supplier No. A13113.AB
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1066-35-9 | C2H6ClSi | 93.61 g/mol

Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

Solubility
Miscible with organic solvents.

Notes
It is incompatible with strong oxidizing agents, strong acids, strong bases, metals and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1066-35-9
Molecular Formula C2H6ClSi
Molecular Weight (g/mol) 93.61
MDL Number MFCD00000495
InChI Key QABCGOSYZHCPGN-UHFFFAOYSA-N
Synonym chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical
PubChem CID 6327132
IUPAC Name chloro(dimethyl)silicon
SMILES C[Si](C)Cl

Specifications

Boiling Point 34°C to 36°C
Melting Point -111°C
Quantity 10 mL
Linear Formula (CH3)2SiHCl
UN Number UN2924
Beilstein 1633434
Flash Point −28°C (−18°F)
Formula Weight 94.62
Percent Purity 97%
Odor Acrid
Chemical Name or Material Chlorodimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.