missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine, 95%, tech.

Product Code. 10637364 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10637364 500 mL Glass bottle 500mL
10071020 25 mL Glass bottle 25mL
10081020 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10637364 Supplier Thermo Scientific Supplier No. 109135000
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
SMILES ClP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.2290g/mL
Boiling Point 320.0°C
Flash Point 138°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2PCl
Refractive Index 1.6340 to 1.6370
Quantity 500 mL
Beilstein 16, 763
Fieser 11,120; 16,77
Merck Index 15, 2139
Specific Gravity 1.229
Solubility Information Solubility in water: reacts violently with water
IUPAC Name chloro(diphenyl)phosphane
Formula Weight 220.64
Percent Purity 95%
Grade Technical
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chlorodiphenylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H290-May be corrosive to metals.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.