missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ≥99.5%, Honeywell™

Product Code. 15687730 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
4 x 25 mL
500 mL
2.5 L
6 x 500 mL
Packaging:
Glass bottle
Unit Size:
2.5 L
25mL
500mL
Pack of 4
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15687730 500 mL Glass bottle 500mL
15668480 25 mL Glass bottle 25mL
15677730 4 x 25 mL Glass bottle Pack of 4
15667730 2.5 L Glass bottle 2.5 L
15697730 6 x 500 mL Glass bottle Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 15687730 Supplier Honeywell Research Chemicals Supplier No. C2432500ML
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Contains 100-200 ppm amylenes as stabilizer,≥99.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -61.3°C
Color Colorless
Density 1.492g/mL at 25°C
Boiling Point 61°C (1013 hPa)
Odor Sweet
Packaging Glass bottle
Quantity 500 mL
UN Number UN1888
Beilstein 1731042
IUPAC Name chloroform
Stabilizer Amylenes
Vapor Density 4.1 (vs air)
Formula Weight 119.38g/mol
Vapor Pressure 160mmHg (20°C)
Percent Purity ≥99.5%
Physical Form Liquid
Chemical Name or Material Chloroform
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.