missing translation for 'onlineSavingsMsg'
Learn More

Chloroform-d, for NMR, 99.8+ atom % D, contains 0.02-0.04 v/v% TMS

Product Code. 10348680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass Bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10348680 100 mL Glass Bottle 100mL
10021621 25 mL Glass Bottle 25mL
10450101 500 mL Glass Bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10348680 Supplier Thermo Scientific Acros Supplier No. 209561000
  • ONLINE EXCLUSIVE PRICE
    91.60 €
    88.90 € / 100mL
    Save 2.70 €
    2% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 865-49-6 | CHCl3 | 120.375 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.375
MDL Number MFCD00000827
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
Synonym chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
PubChem CID 71583
ChEBI CHEBI:85365
IUPAC Name trichloro(deuterio)methane
SMILES C(Cl)(Cl)Cl

Specifications

Physical Form Crystalline Powder
Color White to Beige
Density 1.5000g/mL
Deuterium (D) 99.80% min.
Packaging Glass Bottle
Water 0.01% max. (Coulometric)
Quantity 100 mL
Melting Point -64.0°C
Boiling Point 60.0°C
Linear Formula CDCl3
Beilstein 01, IV, 54
Formula Weight 120.39
Grade NMR
Chemical Name or Material Chloroform-d, 99.8+ atom % D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform-d
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.