missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0-99.4% (GC), Honeywell Riedel-de Haën™
BIOPHARMA

Catalog Number 15675705
104.32 EUR valid until 2024-05-31
Use promo code "21688" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
67-66-3, 64-17-5
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl
67-66-3, 64-17-5
CHCl3
119.37
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
trichloromethane
ClC(Cl)Cl

Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0-99.4% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Melting Point -63°C (literature)
CAS Min % ≥90.0000%, ≥0.1000%
CAS Max % ≤100.0000%, <1.0000%
Density 1.476-1.483 g/mL at 20 °C; 1.492 g/mL at 25 °C(lit.)
Boiling Point 60°C to 62°C
Quantity 2.5 L
Packaging Glass Bottle
Linear Formula CHCl3
UN Number UN1888
Beilstein 1731042
Grade puriss. p.a.
Chemical Name or Material Chloroform, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., 99.0-99.4% (GC)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloroform
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'