missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlorogenic acid, 98%

Product Code. 10349283 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10349283

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 327-97-9 | C16H18O9 | 354.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-97-9
Molecular Formula C16H18O9
Molecular Weight (g/mol) 354.31
MDL Number MFCD00003862
InChI Key CWVRJTMFETXNAD-JUHZACGLSA-N
Synonym chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri
PubChem CID 1794427
ChEBI CHEBI:16112
IUPAC Name (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O

Specifications

Melting Point 205.0°C to 209.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 10, I, 271
Merck Index 15, 2143
Specific Rotation -36
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and acetone
Formula Weight 354.3
Percent Purity 98%
Physical Form Crystals, Powder, Needles or Chunks
Chemical Name or Material Chlorogenic acid, Predominantly trans, From coffee seeds
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.