missing translation for 'onlineSavingsMsg'
Learn More

Chlorogenic Acid, MP Biomedicals™

Product Code. 11457921 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11457921 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11457921 Supplier MP Biomedicals Supplier No. 02150618.1
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chlorogenic Acid is a phenolic product isolated from coffee beans with antimicrobial properties.

Chlorogenic acid is an important factor in plant metabolism.

  • Acts as an antioxidant and an inhibitor of the tumor-promoting activity of phorbol esters
  • At concentrations as high as 100μm, chlorogenic acid does not inhibit the 5-lipoxygenase activity of ionophore-stimulating human polymorphonuclear leukocytes

Specifications

CAS 327-97-9
Melting Point 205°C to 215°C
Density 1.28g/cm3 (Lit.)
Molecular Formula C16H18O9
MDL Number MFCD00003862
Quantity 100 mg
Synonym chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri
Solubility Information Soluble in water or ethanol (25mg/mL, clear to hazy yellow solution).
InChI Key CWVRJTMFETXNAD-JUHZACGLSA-N
SMILES O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O
IUPAC Name (1S,3R,4R,5R)-3-{[(2E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy}-1,4,5-trihydroxycyclohexane-1-carboxylic acid
Molecular Weight (g/mol) 354.31
PubChem CID 1794427
ChEBI CHEBI:16112
Formula Weight 354.3
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.