missing translation for 'onlineSavingsMsg'
Learn More

Chloromethanesulfonyl chloride, 95%

Product Code. 12367093 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12367093 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12367093 Supplier Thermo Scientific Acros Supplier No. 222460050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3518-65-8 | CH2Cl2O2S | 148.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3518-65-8
Color White
Density 1.6500g/mL
Boiling Point 80.0°C to 81.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Molecular Formula CH2Cl2O2S
Linear Formula ClCH2SO2Cl
MDL Number MFCD00014727
Quantity 5 g
Beilstein 01,III,2592
Specific Gravity 1.65
Synonym methanesulfonyl chloride, chloro, chloromethylsulfonyl chloride, chloromethanesulphonyl chloride, chlormethansulfochlorid, chlormethansulfonylchlorid, chloromesyl chloride, chloromethyl sulfonyl chloride, chlormethansulfochlorid czech, chlormethansulfonylchlorid czech, chlorid kyseliny chlormethansulfonove
Solubility Information Solubility in water: reacts
InChI Key KQDDQXNVESLJNO-UHFFFAOYSA-N
SMILES ClCS(Cl)(=O)=O
IUPAC Name chloromethanesulfonyl chloride
Molecular Weight (g/mol) 148.99
PubChem CID 77054
Formula Weight 149
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Chloromethanesulfonyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloromethanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.