missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl chloroformate, 97%

Product Code. 11327476
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327476 50 g 50g
11317476 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327476 Supplier Thermo Scientific Alfa Aesar Supplier No. L09527.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22128-62-7 | C2H2Cl2O2 | 128.936 g/mol

Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22128-62-7
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00077688
InChI Key JYWJULGYGOLCGW-UHFFFAOYSA-N
Synonym chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
PubChem CID 62754
IUPAC Name chloromethyl carbonochloridate
SMILES C(OC(=O)Cl)Cl

Specifications

Melting Point <-20°C
Density 1.465
Boiling Point 107°C to 108°C
Flash Point 95°C (203°F)
Odor Acrid
Linear Formula ClCO2CH2Cl
Refractive Index 1.4295
Quantity 50 g
UN Number UN2745
Beilstein 506426
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water.
Formula Weight 128.94
Percent Purity 97%
Chemical Name or Material Chloromethyl chloroformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloromethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Chloromethylchloroformate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.