missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)cyclopropane, 97%

Product Code. 11367076
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367076 25 g 25g
11357076 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Codice prodotto. 11367076 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L06986.14
 Restricted Item
Estimated Shipment: 30-12-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 5911-08-0 | C4H7Cl | 90.55 g/mol

(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4?-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)cyclopropane was used in the synthesis of (lithiomethyl)cyclopropane and homoallyllithium via 4,4′-di-tert-butylbiphenyl or naphthalene-catalysed lithiation reaction. On reaction with isolable dialkylsilylene affords 5-silaspiro[4.4]nonane derivative.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, bases and heat.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 5911-08-0
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00001307
InChI Key ZVTQWXCKQTUVPY-UHFFFAOYSA-N
Synonym chloromethyl cyclopropane, cylopropylmethyl chloride, cyclopropylmethyl chloride, cyclopropane, chloromethyl, chloromethyl-cyclopropane, j7okqcc0kw, unii-j7okqcc0kw, cylopropylmethylchloride, chlorocyclopropylmethane, cyclopropylchloromethane
PubChem CID 80013
SMILES ClCC1CC1

Specifica

Melting Point -91°C
Density 0.98
Boiling Point 87°C to 89°C
Flash Point −1°C (30°F)
Odor Recognizable
Refractive Index 1.436
Quantity 25 g
UN Number UN1993
Beilstein 635697
Solubility Information Not miscible in water.
IUPAC Name (chloromethyl)cyclopropane
Formula Weight 90.55
Percent Purity 97%
Chemical Name or Material (Chloromethyl)cyclopropane
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.