missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl pivalate, 97%

Product Code. 11473984
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473984 25 g 25g
11483984 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11473984 Supplier Thermo Scientific Alfa Aesar Supplier No. A11967.14
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 18997-19-8 | C6H11ClO2 | 150.60 g/mol

Chloromethyl pivalate acts as an intermediate in the synthesis of active pharmaceutical ingredients. It is also involved in the acylation reaction with 9-(2-phosphonylmethoxyethyl)adenine. It serves as a protecting reagent used for the N-protection of amines. It is also employed in the preparation of pivaloyloxy methyl ester of ofloxacin as a prodrug. Further, it is used to prepare sulbactam pivoxil by reaction with sodium salt of sulbactam. In addition to this, it is used as a reagent during the synthesis of an isoindoline-annulated and continuous-flow organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloromethyl pivalate acts as an intermediate in the synthesis of active pharmaceutical ingredients. It is also involved in the acylation reaction with 9-(2-phosphonylmethoxyethyl)adenine. It serves as a protecting reagent used for the N-protection of amines. It is also employed in the preparation of pivaloyloxy methyl ester of ofloxacin as a prodrug. Further, it is used to prepare sulbactam pivoxil by reaction with sodium salt of sulbactam. In addition to this, it is used as a reagent during the synthesis of an isoindoline-annulated and continuous-flow organic synthesis.

Solubility
Miscible with most organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18997-19-8
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00000884
InChI Key GGRHYQCXXYLUTL-UHFFFAOYSA-N
Synonym chloromethyl pivalate, propanoic acid, 2,2-dimethyl-, chloromethyl ester, pivalic acid chloromethyl ester, pivaloyloxymethyl chloride, pivaloylmethyl chloride, chloromethyl trimethylacetate, 2,2-dimethylpropionic acid chloromethyl ester, chioromethyl pivalate, chloromethyl pivaloate, chloro-methyl pivalate
PubChem CID 87885
IUPAC Name chloromethyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCCl

Specifications

Density 1.045
Boiling Point 146°C to 148°C
Flash Point 40°C (104°F)
Linear Formula (CH3)3CCO2CH2Cl
Refractive Index 1.417
Quantity 25 g
UN Number UN2924
Beilstein 1560838
Solubility Information Miscible with most organic solvents. Immiscible with water.
Formula Weight 150.6
Percent Purity 97%
Chemical Name or Material Chloromethyl pivalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.