missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl pivalate, 99+%

Product Code. 10711925 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10711925 500 g Glass bottle 500g
10468710 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711925 Supplier Thermo Scientific Acros Supplier No. 161465000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 18997-19-8 | C6H11ClO2 | 150.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18997-19-8
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00000884
InChI Key GGRHYQCXXYLUTL-UHFFFAOYSA-N
Synonym chloromethyl pivalate, propanoic acid, 2,2-dimethyl-, chloromethyl ester, pivalic acid chloromethyl ester, pivaloyloxymethyl chloride, pivaloylmethyl chloride, chloromethyl trimethylacetate, 2,2-dimethylpropionic acid chloromethyl ester, chioromethyl pivalate, chloromethyl pivaloate, chloro-methyl pivalate
PubChem CID 87885
IUPAC Name chloromethyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCCl

Specifications

Color Colorless to Yellow
Density 1.0380g/mL
Boiling Point 146°C to 148°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CCO2CH2Cl
Refractive Index 1.4160 to 1.418
Quantity 500 g
Specific Gravity 1.038
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,toluene and chlorinated,hydrocarbons
Formula Weight 150.6
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Chloromethyl pivalate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Chloromethyl pivalate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.