missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethyl)trimethylsilane, 98%

Product Code. 11456837
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456837 250 g 250g
11432487 10 g 10g
11442487 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456837 Supplier Thermo Scientific Alfa Aesar Supplier No. A14303.30
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2344-80-1 | C4H11ClSi | 122.667 g/mol

(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethyl)trimethylsilane is used for Wittig reaction and building blocks.

Solubility
Soluble in water (0.4g/L at 20°C).

Notes
Store in cool, well ventilated area. Keep container tightly closed. Moisture sensitive. Store under argon.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2344-80-1
Molecular Formula C4H11ClSi
Molecular Weight (g/mol) 122.667
MDL Number MFCD00000878
InChI Key OOCUOKHIVGWCTJ-UHFFFAOYSA-N
Synonym chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane
PubChem CID 75361
IUPAC Name chloromethyl(trimethyl)silane
SMILES C[Si](C)(C)CCl

Specifications

Boiling Point 97°C to 98°C
Quantity 250 g
UN Number UN1993
Beilstein 906705
Flash Point −2°C (28°F)
Solubility Information Soluble in water (0.4g/L at 20°C).
Formula Weight 122.67
Percent Purity 98%
Odor Odorless
Refractive Index 1.418
Sensitivity Moisture sensitive
Density 0.886
Chemical Name or Material (Chloromethyl)trimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.