missing translation for 'onlineSavingsMsg'
Learn More

(Chloromethylene)dimethylammonium Chloride, 96%

Product Code. 11411207
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411207 100 g 100g
11491197 5 g 5g
11401207 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11411207 Supplier Thermo Scientific Alfa Aesar Supplier No. B24172.22
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 3724-43-4 | C3H7Cl2N | 127.996 g/mol

(Chloromethylene)dimethylammonium chloride is used in the synthesis of beta-lactams, n-vinyl substituted indoles and fused 2-pyridone rings and 2-azetidinones. It is used as a formylating agent and used in the preparation of anti-poxvirus agents. It is used in Staudinger reaction and Hundsdiecker-barton iododecarboxylation reaction. It is widely used for studying organic photosensitizers, which finds application in dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Chloromethylene)dimethylammonium chloride is used in the synthesis of beta-lactams, n-vinyl substituted indoles and fused 2-pyridone rings and 2-azetidinones. It is used as a formylating agent and used in the preparation of anti-poxvirus agents. It is used in Staudinger reaction and Hundsdiecker-barton iododecarboxylation reaction. It is widely used for studying organic photosensitizers, which finds application in dye-sensitized solar cells.

Solubility
Soluble in water.

Notes
Moisture sensitive. It may decompose in moist air/ water. Incompatible with bases, strong oxidizing agents, amines, metals and butyl rubber.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3724-43-4
Molecular Formula C3H7Cl2N
MDL Number MFCD00011868
InChI Key QQVDYSUDFZZPSU-UHFFFAOYSA-M
PubChem CID 77311
IUPAC Name chloromethylidene(dimethyl)azanium;chloride

Specifications

Melting Point 130°C (decomposition)
Odor Acrid
Quantity 100 g
UN Number UN3261
Beilstein 3566322
Sensitivity Moisture sensitive
Synonym arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride
Solubility Information Soluble aq. soln..
SMILES [Cl-].CN(C)[CH+]Cl
Molecular Weight (g/mol) 128.00
Formula Weight 128
Percent Purity 96%
Chemical Name or Material (Chloromethylene)dimethylammonium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.