missing translation for 'onlineSavingsMsg'
Learn More

Chloropentaammineiridium(III) chloride, 99.9% (metals basis), Ir 49.6% min

Product Code. 11389397
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
This item is not returnable. View return policy

Product Code. 11389397

Brand: Thermo Scientific Alfa Aesar 041006.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15742-38-8 | Cl3H15IrN5 | 383.722 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15742-38-8
Molecular Formula Cl3H15IrN5
Molecular Weight (g/mol) 383.722
MDL Number MFCD00798542
InChI Key DYGMZANLQHDDSH-UHFFFAOYSA-K
Synonym chloropentaammineiridium iii chloride, cl3ir.5nh3, chloropentaammineiridium iii chloride, ir min, pentaamminechloroiridium iii chloride trace metals basis
PubChem CID 15541975
IUPAC Name azane;trichloroiridium
SMILES N.N.N.N.N.Cl[Ir](Cl)Cl

Specifications

Melting Point (decomposition)
Color White to Yellow
Linear Formula [IrCl(NH3)5]Cl2
Quantity 1 g
Formula Weight 383.73
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Ir 49.6% min.
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Chloropentaammineiridium(III) chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.