missing translation for 'onlineSavingsMsg'
Learn More

Chloroplatinic Acid Hexahydrate, Honeywell™

Product Code. 15688040 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
15688040 5 g Glass bottle 5g
1 options
This item is not returnable. View return policy

Product Code. 15688040

Brand: Honeywell-Fluka P57755G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Powder and chunks

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 18497-13-7
Color Orange
Quantity 5 g
Molecular Formula Cl6H14O6Pt
Linear Formula H2PtCl6 · 6H2O
MDL Number MFCD00149910
UN Number UN2507
InChI Key PIJUVEPNGATXOD-UHFFFAOYSA-J
SMILES [H+].[H+].O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Pt+4]
IUPAC Name platinum(4+) dihydrogen hexahydrate hexachloride
Molecular Weight (g/mol) 517.89
Formula Weight 517.9g/mol
Packaging Glass bottle
Chemical Name or Material Chloroplatinic acid hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.