missing translation for 'onlineSavingsMsg'
Learn More

Chloropyramine hydrochloride, MedChemExpress

Product Code. 16494577
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16494577 Supplier Medchem Express Supplier No. HYB1305
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chloropyramine hydrochloride is a histamine receptor H1 antagonist which can also inhibit the biochemical function of VEGFR-3 and FAK.

Chloropyramine hydrochloride is a histamine receptor H1 antagonist which can also inhibit the biochemical function of VEGFR-3 and FAK.

Specifications

Chemical Name or Material Chloropyramine hydrochloride
CAS 6170-42-9
Color White
Purity Grade Notes Research
Molecular Formula C16H21Cl2N3
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (306.50 mM; Need ultrasonic) ∣DMSO : 30 mg/mL (91.95 mM; Need ultrasonic and warming)
SMILES CN(C)CCN(CC1=CC=C(Cl)C=C1)C2=NC=CC=C2.[H]Cl
Molecular Weight (g/mol) 326.26
Formula Weight 326.26
Percent Purity 99.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.