missing translation for 'onlineSavingsMsg'
Learn More

Chloroquine Diphosphate Salt, TRC

Product Code. 17934815
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17934815 100 mg 100mg
17912575 10 mg 10mg
17902675 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17934815 Supplier Toronto Research Chemicals Supplier No. TRCC379965100MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18 H26 Cl N3 . 2 H3 O4 P
Molecular Weight (g/mol) 515.86
Synonym Quinoline, 7-chloro-4-[(4-diethylamino-1-methylbutyl)amino]-, diphosphate (6CI), Quinoline, 7-chloro-4-[[4-(diethylamino)-1-methylbutyl]amino]-, phosphate (1:2) (8CI), (±)-Chloroquine diphosphate, 3377RP, 7-Chloro-4-[(4'-diethylamino-1-methylbutyl)amino]quinoline diphosphate, Alexoquine, Aralen diphosphate, Aralen phosphate, Arechin, Avloclor, Bemaphate, Chingamin, Chingamin phosphate, Chlorochin diphosphate, Chloroquin diphosphate, Chloroquine dihydrogen phosphate (1:2), Chloroquine diphosphate, Chloroquine phosphate, Cidoquine, Delagil, Gontochin phosphate, Imagon, Khingamin, Maladrin, Malaquin, NSC 14050, Nivaquine B, Quingamine, Quivaclor, Resochin, Resoquine, SN 7618, Sanoquin, Tanakan, Tanakan (antimalarial), Tresochin, WR 1522, dl-Chloroquine diphosphate
IUPAC Name 4-N-(7-chloroquinolin-4-yl)-1-N,1-N-diethylpentane-1,4-diamine;phosphoric acid
SMILES CCN(CC)CCCC(C)Nc1ccnc2cc(Cl)ccc12.OP(=O)(O)O.OP(=O)(O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C18H26ClN3.2H3O4P/c1-4-22(5-2)12-6-7-14(3)21-17-10-11-20-18-13-15(19)8-9-16(17)18;2*1-5(2,3)4/h8-11,13-14H,4-7,12H2,1-3H3,(H,20,21);2*(H3,1,2,3,4)
Formula Weight 515.1353
Chemical Name or Material Chloroquine Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.