missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Chloroquine diphosphate

Product Code. 18762474
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18762474 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18762474 Supplier R&D Systems™ Supplier No. 4109/50
  • ONLINE EXCLUSIVE PRICE
    99.96 €
    89.25 € / 50mg
    Save 10.71 €
    10% Off
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Antimalarial; inhibits apoptosis and autophagy

Chloroquine diphosphate is an antimalarial drug. Inhibits cell growth and induces cell death in numerous cancer cell lines; inhibits cell proliferation and viability and induces apoptosis in 4T1 mouse breast cancer cells in vitro. Exhibits antimetastatic activity. Also inhibits autophagy via a mechanism distinct from that of 3-methyladenine (Cat. No. 3977). Blocks receptor-mediated endocytosis of mannose-glycoconjugates by macrophages. Inhibits SARS-CoV-2 infection in vitro (EC50 = 1.13 μM). Chloroquine improves efficacy of adeno-associated viral gene transduction in vivo and in vitro, as well as enhancing non-viral gene transfection in breast cancer cells. R&D Systems products are for biomedical research use only. They are not intended for human or veterinary use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 50-63-5
Quantity 50 mg
Product Line Tocris
Molecular Formula C18H32ClN3O8P2
Formula Weight 515.86
Purity >99%
Molecular Weight (g/mol) 515.86
Chemical Name or Material Chloroquine diphosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.