missing translation for 'onlineSavingsMsg'
Learn More

Chlorothiazide-13C,15N2, TRC

Product Code. 30133221
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133221 5 mg 5mg
30011624 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133221 Supplier Toronto Research Chemicals Supplier No. TRCC3800025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189440-79-6
Molecular Formula C613CH6ClN15N2O4S2
Molecular Weight (g/mol) 298.7
Synonym 6-Chloro-7-sulfamyl-1,2,4-benzothiadiazine-1,1-dioxide-13C,15N2, 6-Chloro-2H-1,2,4-benzothiadiazine-7-sulfonamide-13C,15N2 1,1-dioxide, Chlorthiazide-13C,15N2;Chlotride-13C,15N2;Diuril-13C,15N2, Saluric-13C,15N2;
IUPAC Name 6-chloro-4H-benzo[e][1,2,4]thiadiazine-7-sulfonamide-3-13C-2-15N-15N 1,1-dioxide
SMILES ClC1=C(S([15NH2])(=O)=O)C=C2C(N[13CH]=[15N]S2(=O)=O)=C1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C7H6ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-3H,(H,10,11)(H2,9,12,13)/i3+1,9+1,11+1
Formula Weight 297.95
Percent Purity >95%
Chemical Name or Material Chlorothiazide-13C,15N2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.