missing translation for 'onlineSavingsMsg'
Learn More

Chlorothiazide, 98%

Product Code. 15703457 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15703457 100 g 100g
15783447 5 g 5g
15793447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15703457 Supplier Thermo Scientific Acros Supplier No. 461741000
 Restricted Item
In Stock
Add to basket
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 58-94-6 | C7H6ClN3O4S2 | 295.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 58-94-6
Molecular Formula C7H6ClN3O4S2
Molecular Weight (g/mol) 295.71
MDL Number MFCD00058576
InChI Key JBMKAUGHUNFTOL-UHFFFAOYSA-N
Synonym chlorothiazide, diuril, chlorthiazide, chlorothiazid, chlotride, chlortiazid, thiazide, flumen, chlorosal, chlorurit
PubChem CID 2720
ChEBI CHEBI:3640
IUPAC Name 6-chloro-1,1-dioxo-4H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NC=NS2(=O)=O)C=C1Cl

Spécification

Melting Point 348.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Solubility Information Solubility in water: very slightly soluble. Other solubilities: freely soluble in dmso, dmf, slightly soluble in methanol, pyridine, practically insoluble in diethyl ether, benzene, and chloroform
Formula Weight 295.72
Percent Purity 98%
Chemical Name or Material Chlorothiazide

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.