missing translation for 'onlineSavingsMsg'
Learn More

Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. 15441496
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15441496 25 g 25g
15421496 1 g 1g
15431496 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15441496 Supplier Thermo Scientific Alfa Aesar Supplier No. H54693.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133894-48-1
Molecular Formula C12H24ClF6N3P2
Molecular Weight (g/mol) 421.733
MDL Number MFCD00210035
InChI Key BSCYRXJVGSZNKX-UHFFFAOYSA-N
Synonym pyclop, chlorotripyrrolidinophosphonium hexafluorophosphate, chlorotripyrrolidinophosphonium hexa-fluorophosphate, chlorotri 1-pyrrolidinyl phosphonium hexafluorophosphate, 98+%, pyciop, ref dupl: pyclop, chlorotripyrrolidino, acmc-1c01w, ksc177a1n, chlorotripyrrolidinophosphoniumhexafluorophosphat
PubChem CID 16211753
IUPAC Name chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
SMILES C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F

Specifications

Melting Point 145°C
Quantity 25 g
UN Number UN1759
Beilstein 6842341
Sensitivity Moisture sensitive
Formula Weight 421.73
Percent Purity ≥98%
Chemical Name or Material Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.