missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriethylsilane, 98+%

Product Code. 11482977
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11482977

Brand: Thermo Scientific Alfa Aesar A15547.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 994-30-9 | C6H15ClSi | 150.72 g/mol

Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriethylsilane is used in laboratory chemicals. It is used for preparing silylating agent and acts as a catalyst for lewis acid. It is used for executing chemical reactions for silicon oil, silicon resin and polysiloxane. It is also used for eradicating robe agent, and used as protective agent and leather surface treating agent.

Solubility
Soluble in aprotic organic solvents.

Notes
Incompatible with bases, strong oxidizing agents, strong acids, alcohols, amines, ammonia, aldehydes and acetone.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-30-9
Molecular Formula C6H15ClSi
Molecular Weight (g/mol) 150.72
MDL Number MFCD00000507
InChI Key DCFKHNIGBAHNSS-UHFFFAOYSA-N
Synonym triethylchlorosilane, silane, chlorotriethyl, triethylsilyl chloride, chloro triethyl silane, unii-q1a687m1py, triethyl silyl chloride, triethychlorosilane, chlorotrietylsilane, clsiet3, et3sicl
PubChem CID 13819
IUPAC Name chloro(triethyl)silane
SMILES CC[Si](Cl)(CC)CC

Specifications

Boiling Point 145°C to 147°C
Melting Point ∼-75°C
Quantity 10 g
Linear Formula (CH3CH2)3SiCl
UN Number UN2986
Beilstein 1732860
Flash Point 29°C (84°F)
Solubility Information Soluble in aprotic organic solvents.
Formula Weight 150.73
Percent Purity ≥98%
Odor Pungent
Refractive Index 1.431
Sensitivity Moisture sensitive
Density 0.899
Chemical Name or Material Chlorotriethylsilane
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorotriethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Chlorotriethylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.