missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriisopropylsilane, 97+%

Product Code. 11483617
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11483617

Brand: Thermo Scientific Alfa Aesar A17376.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13154-24-0 | C9H21ClSi | 192.80 g/mol

Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotriisopropylsilane is used as a silylating agent in neucleotide synthesis. It is used as an intermediate in the manufacture of chemical substances such as pharmaceuticals and in organic synthesis.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13154-24-0
Molecular Formula C9H21ClSi
Molecular Weight (g/mol) 192.80
MDL Number MFCD00009656
InChI Key KQIADDMXRMTWHZ-UHFFFAOYSA-N
Synonym triisopropylsilyl chloride, chlorotriisopropylsilane, triisopropylchlorosilane, chlorotris propan-2-yl silane, chloro triisopropyl silane, silane, chlorotris 1-methylethyl, tipscl, triisopropylsilylchloride, triisopropyl chlorosilane, triisopropylchlorosilane, redistilled
PubChem CID 139400
IUPAC Name chloro-tri(propan-2-yl)silane
SMILES CC(C)[Si](Cl)(C(C)C)C(C)C

Specifications

Boiling Point 200°C
Quantity 10 g
UN Number UN2987
Beilstein 1737446
Flash Point 62°C (144°F)
Solubility Information Miscible with water.
Formula Weight 192.81
Percent Purity ≥97%
Odor Acrid
Refractive Index 1.4535
Sensitivity Moisture sensitive
Density 0.908
Chemical Name or Material Chlorotriisopropylsilane
Show More Show Less
compliance-icons
Product Identifier
  • Chlorotriisopropylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Chlorotriisopropylsilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.