missing translation for 'onlineSavingsMsg'
Learn More

chlorotrimethylsilane, 1M Solution in Dichloromethane, AcroSeal™, ACROS Organics™

Product Code. 10482863 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100mL; AcroSeal glass bottle packed in tin can
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10482863 100mL; AcroSeal glass bottle packed in tin can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482863 Supplier Thermo Scientific Acros Supplier No. 381601000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Silylation reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 75-77-4, 75-09-2
Density 1.2570g/mL
Packaging AcroSeal™ Glass Bottle in Tin Can
Molecular Formula C3H9ClSi
Linear Formula (CH3)3SiCl
MDL Number MFCD00000502
Quantity 100 mL
Merck Index 15, 2180
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmscl
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
SMILES C[Si](C)(C)Cl
IUPAC Name chloro(trimethyl)silane
Molecular Weight (g/mol) 108.64
PubChem CID 6397
ChEBI CHEBI:85069
Formula Weight 108.64
Chemical Name or Material Chlorotrimethylsilane, 1M Solution in Dichloromethane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.