missing translation for 'onlineSavingsMsg'
Learn More

Chlorotriphenylsilane, 95%

Product Code. 10498490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498490 100 g Glass bottle 100g
10408690 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10498490 Supplier Thermo Scientific Acros Supplier No. 151251000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76-86-8 | C18H15ClSi | 294.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-86-8
Molecular Formula C18H15ClSi
Molecular Weight (g/mol) 294.85
MDL Number MFCD00000496
InChI Key MNKYQPOFRKPUAE-UHFFFAOYSA-N
Synonym triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl
PubChem CID 6458
IUPAC Name chloro(triphenyl)silane
SMILES Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 91°C to 96°C
Color White to Beige
Boiling Point 378°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)3SiCl
Quantity 100 g
Beilstein 16, 905
Fieser 12,279
Solubility Information Solubility in water: decomposes
Formula Weight 294.85
Percent Purity 95%
Physical Form Crystalline Powder and Crystals
Chemical Name or Material Chlorotriphenylsilane, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Chlorotriphenylsilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.