missing translation for 'onlineSavingsMsg'
Learn More

Chlorpromazine Sulfoxide, TRC

Product Code. 30007351
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007351 25 mg 25mg
30006251 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007351 Supplier Toronto Research Chemicals Supplier No. TRCC42475525MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 969-99-3
Molecular Formula C17 H19 Cl N2 O S
Molecular Weight (g/mol) 334.86
Synonym 10H-Phenothiazine-10-propanamine, 2-chloro-N,N-dimethyl-, 5-oxide, Phenothiazine, 2-chloro-10-[3-(dimethylamino)propyl]-, 5-oxide (6CI,8CI), Aminazin sulfoxide, Ba 2835, Chlorpromazine 5-oxide, Chlorpromazine 5-sulfoxide, Chlorpromazine S-oxide, Chlorpromazine sulfoxide, Chlorpromazine sulphoxide, NSC 170990, Opromazine, 3-(2-Chloro-10H-phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide
IUPAC Name 3-(2-chloro-5-oxophenothiazin-10-yl)-N,N-dimethylpropan-1-amine
SMILES CN(C)CCCN1c2ccccc2S(=O)c3ccc(Cl)cc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H19ClN2OS/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)22(21)17-9-8-13(18)12-15(17)20/h3-4,6-9,12H,5,10-11H2,1-2H3
Formula Weight 334.0907
Percent Purity >95%
Chemical Name or Material Chlorpromazine Sulfoxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.