missing translation for 'onlineSavingsMsg'
Learn More

Chlorprothixene Hydrochloride, TRC

Product Code. 30041263
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041263 5 mg 5mg
30040958 100 mg 100mg
18090020 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041263 Supplier Toronto Research Chemicals Supplier No. TRCC4248505MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6469-93-8
Molecular Formula C18 H18 Cl N S . Cl H
Molecular Weight (g/mol) 352.32
Synonym Chlorprothixene hydrochloride, (Z)-3-(2-Chloro-9H-thioxanthen-9-ylidene)-N,N-dimethylpropan-1-amine hydrochloride, 1-Propanamine, 3-(2-chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-, hydrochloride (1:1), (3Z)-, 1-Propanamine, 3-(2-chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-, hydrochloride, (3Z)- (9CI), 1-Propanamine, 3-(2-chloro-9H-thioxanthen-9-ylidene)-N,N-dimethyl-, hydrochloride, (Z)-, Thioxanthene-δ9,γ-propylamine, 2-chloro-N,N-dimethyl-, hydrochloride, (Z)- (8CI), 9H-Thioxanthene, 1-propanamine deriv., NSC 169899, Truxal hydrochloride, cis-Chlorprothixene hydrochloride
IUPAC Name (3Z)-3-(2-chlorothioxanthen-9-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
SMILES Cl.CN(C)CC\C=C/1\c2ccccc2Sc3ccc(Cl)cc13

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H18ClNS.ClH/c1-20(2)11-5-7-14-15-6-3-4-8-17(15)21-18-10-9-13(19)12-16(14)18;/h3-4,6-10,12H,5,11H2,1-2H3;1H/b14-7-;
Formula Weight 351.0615
Chemical Name or Material Chlorprothixene hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.