missing translation for 'onlineSavingsMsg'
Learn More

Chlorsulfonyl Isocyanate, TRC

Product Code. 30359656
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30359656 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30359656

Brand: Toronto Research Chemicals TRCC38018510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorsulfonyl Isocyanate
CAS 1189-71-5
Molecular Formula CClNO3S
Quantity 10 g
Synonym Sulfuryl Chloride Isocyanate, Isocyanic Acid Anhydride with Chlorosulfonic Acid, CSI, Chlorosulfuric Acid Anhydride with Isocyanic Acid, Chlorosulphonyl Isocyanate, Chlorsulfonyl Isocyanate, N-Chlorosulfonyl Isocyanate, N-Chlorosulphonyl Isocyanate, Sulfonyl Chloride Isocyanate
InChI Formula InChI=1S/CClNO3S/c2-7(5,6)3-1-4
SMILES ClS(=O)(=O)N=C=O
IUPAC Name N-(oxomethylidene)sulfamoyl chloride
Molecular Weight (g/mol) 141.53
Formula Weight 140.9287
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.