missing translation for 'onlineSavingsMsg'
Learn More

Chlorsulfonyl Isocyanate, TRC

Product Code. 30358897
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity unitSize
30358897 50 g 50g
1 options
This item is not returnable. View return policy

Product Code. 30358897

Brand: Toronto Research Chemicals TRCC38018550G

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Chlorsulfonyl Isocyanate
CAS 1189-71-5
Molecular Formula CClNO3S
Quantity 50 g
Synonym Sulfuryl Chloride Isocyanate, Isocyanic Acid Anhydride with Chlorosulfonic Acid, CSI, Chlorosulfuric Acid Anhydride with Isocyanic Acid, Chlorosulphonyl Isocyanate, Chlorsulfonyl Isocyanate, N-Chlorosulfonyl Isocyanate, N-Chlorosulphonyl Isocyanate, Sulfonyl Chloride Isocyanate
InChI Formula InChI=1S/CClNO3S/c2-7(5,6)3-1-4
SMILES ClS(=O)(=O)N=C=O
IUPAC Name N-(oxomethylidene)sulfamoyl chloride
Molecular Weight (g/mol) 141.53
Formula Weight 140.9287
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.