missing translation for 'onlineSavingsMsg'
Learn More

Chlorthalidone, TRC

Product Code. 17991865
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991865 10 mg 10mg
17983145 5 mg 5mg
17943835 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17991865 Supplier Toronto Research Chemicals Supplier No. TRCC42750010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-36-1
Molecular Formula C14 H11 Cl N2 O4 S
Molecular Weight (g/mol) 338.77
Synonym Benzenesulfonamide, 2-chloro-5-(2,3-dihydro-1-hydroxy-3-oxo-1H-isoindol-1-yl)-, Benzenesulfonamide, 2-chloro-5-(1-hydroxy-3-oxo-1-isoindolinyl)- (6CI,8CI), 2-Chloro-5-(2,3-dihydro-1-hydroxy-3-oxo-1H-isoindol-1-yl)benzenesulfonamide, (±)-Chlorthalidone, (±)-Hygroton, 1-Oxo-3-(3-sulfamyl-4-chlorophenyl)-3-hydroxy-isoindolinum, 1-Oxo-3-(3-sulfamyl-4-chlorophenyl)-3-hydroxyisoindoline, 1-keto-3-(3'-Sulfamyl-4'-chlorophenyl)-3-hydroxyisoindoline, 2-Chloro-5-(1-hydroxy-3-oxo-1-isoindolinyl)benzenesulfonamide, 2-Chloro-5-(3-hydroxy-1-oxoisoindol-3-yl)benzenesulfonamide, 3-(4'-Chloro-3'-sulfamoylphenyl)-3-hydroxyphthalimidine, 3-Hydroxy-3-[4-chloro-3-sulfamylphenyl]phthalimidine, Chlorothalidone, Chlortalidone, Chlorthalidon, Chlorthalidone, Demi-Regroton, G 33182, Higroton, Higrotona, Hygroton, Igroton, Isoren, NSC 69200, Natriuran, Oradil, Oxodolin, Phthalamudine, Racemic chlorthalidone, Renon, Saluretin, Thalitone, ZINC 00020253, Zambesil
IUPAC Name 2-chloro-5-(1-hydroxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
SMILES NS(=O)(=O)c1cc(ccc1Cl)C2(O)NC(=O)c3ccccc23

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H11ClN2O4S/c15-11-6-5-8(7-12(11)22(16,20)21)14(19)10-4-2-1-3-9(10)13(18)17-14/h1-7,19H,(H,17,18)(H2,16,20,21)
Formula Weight 338.0128
Percent Purity >95%
Chemical Name or Material Chlortalidone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.