missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHMP7 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 462.00 €
Specifications
| Antigen | CHMP7 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
CHMP7 Polyclonal antibody specifically detects CHMP7 in Mouse samples. It is validated for Western BlotSpecifications
| CHMP7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cell Biology, Signal Transduction | |
| PBS with 50% glycerol, pH7.3. | |
| 91782 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| charged multivesicular body protein 7, CHMP family, member 7, Chromatin-modifying protein 7, MGC29816 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 204-453 of human CHMP7 (NP_689485.1). LQKEKRVTVLEQNGEKIVKFARGPRAKVSPVNDVDVGVYQLMQSEQLLSRKVESLSQEAERCKEEARRACRAGKKQLALRSLKAKQRTEKRIEALHAKLDTVQGILDRIYASQTDQMVFNAYQAGVGALKLSMKDVTVEKAESLVDQIQELCDTQDEVSQTLAGGVTNGLDFDSEELEKELDILLQDTTKEPLDLPDNPRNRHFTNSVPNPRISDAELEAELEKLSLSEGGLVPSSKSPKRQLEPTLKPL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title