missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol 5Alpha,6Alpha-Epoxide, TRC

Product Code. 30135964
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135964 100 mg 100mg
30133018 50 mg 50mg
30009096 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135964 Supplier Toronto Research Chemicals Supplier No. TRCC432528100MG
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1250-95-9
Molecular Formula C27 H46 O2
Molecular Weight (g/mol) 402.65
Synonym Cholestan-3-ol, 5,6-epoxy-, (3β,5α,6α)-, 5α-Cholestan-3β-ol, 5,6α-epoxy- (6CI,7CI,8CI), (3β,5α,6α)-5,6-Epoxycholestan-3-ol, 5,6-Epoxy-5H-cyclopenta[a]phenanthrene, cholestan-3-ol deriv., 3β-Hydroxy-5,6α-epoxy-5α-cholestane, 3β-Hydroxycholestane 5α,6α-epoxide, 5,6α-Epoxy-5α-cholestan-3β-ol, 5,6α-Epoxycholesterol, 5α,6α-Epoxycholestan-3β-ol, 5α,6α-Epoxycholestanol, 5α,6α-Epoxycholesterol, 5α-Epoxycholesterol, Cholesterol 5α,6α-epoxide, Cholesterol 5α,6α-oxide, Cholesterol α-epoxide, Cholesterol α-oxide, NSC 18176
IUPAC Name (1S,2R,5S,7R,9S,11S,12S,15R,16R)-2,16-dimethyl-15-[(2R)-6-methylheptan-2-yl]-8-oxapentacyclo[9.7.0.02,7.07,9.012,16]octadecan-5-ol
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3C[C@@H]4O[C@@]45C[C@@H](O)CC[C@]5(C)[C@H]3CC[C@]12C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C27H46O2/c1-17(2)7-6-8-18(3)21-9-10-22-20-15-24-27(29-24)16-19(28)11-14-26(27,5)23(20)12-13-25(21,22)4/h17-24,28H,6-16H2,1-5H3/t18-,19+,20+,21-,22+,23+,24+,25-,26-,27+/m1/s1
Formula Weight 402.3498
Chemical Name or Material Cholesterol 5alpha,6alpha-Epoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.