missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
250X Cholesterol Lipid Concentrate is a protein-free, chemically-defined supplement used for growing NSO and other lipid dependent cell lines to high densities while supporting growth and protein expression without complicating downstream purification. The supplement can be added to 1X media such as CD Hybridoma and filtered with minimal loss of lipids. Custom formulations and packaging configurations as well as supplements used in feeding strategies are available through our Custom Products Services group.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
| Cell Type | Mammalian Cells |
| Concentration | 250 X |
| Culture Type | Mammalian Cell Culture, Insect Cell Culture |
| Form | Liquid |
| Product Type | Cholesterol Lipid Concentrate |
| Sterility | Sterile |
| pH | 6 to 8 |
| Quantity | 100 mL |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
