missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol, MedChemExpress

Product Code. 16492366
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492366 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16492366 Supplier Medchem Express Supplier No. HYN0322

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cholesterol is the major sterol in mammals and is makes up 20-25% of structural component of the plasma membrane.

Plasma membranes are highly permeable to water but relatively impermeable to ions and protons. Cholesterol plays an important role in determining the fluidity and permeability characteristics of the membrane as well as the function of both the transporters and signaling proteins. Cholesterol is also an endogenous estrogen-related receptor α (ERRα) agonist.

Specifications

Chemical Name or Material Cholesterol
CAS 57-88-5
Color White
Purity Grade Notes Research
Molecular Formula C27H46O
Quantity 500 mg
Solubility Information Ethanol : 14.29 mg/mL (36.96 mM; Need ultrasonic)
SMILES O[C@H](C1)CC[C@@]2(C)C1=CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@@H](CCCC(C)C)C
Molecular Weight (g/mol) 386.65
Formula Weight 386.65
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.